missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Barrel with Metal Toggle Joint Closure

Food contact certified plastic Barrel with Metal Toggle Joint Closure. Suitable for a large number of uses (brining, pickling etc.).

Brand:  Gilac™ G178921

 View more versions of this product

Product Code. 11732555

  • 159.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Food contact certified plastic barrel
  • Suitable for a large number of uses (brining, pickling etc.) when transporting and storing liquid or solid food products
  • Global and specific migration test approved
  • Contents are protected from contamination thanks to its watertight lid
  • Handling is easy and can be opened completely
Specifications

Specifications

Food Contact Certified
Food Certified Barrel
26.4 gal.
White
-40°C to 90°C
Round
650 mm
1.6 ft.
492 mm
4.1 kg
Yes
Metal Toggle Joint
100 L
HDPE
1 Ea.
2.1 ft.
1.3 ft.
400 mm
9 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.