Learn More
Emma Safety Footwear BAS Safety Shoes
All purpose safety boot. The outsole repels acid and oil like an umbrella repels water. Strong ankle supporting upper fits the foot snugly and comfortably.
Brand:  Emma Safety Footwear MM70084842
 View more versions of this product
Product Code. 15988841
  • 130.00 € / 1 pair
Estimated Shipment: 11-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
42
Laces
BAS
Leather
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Emma Safety Footwear BAS Safety Shoes > Size: 42