Learn More
Cytiva BAS Storage Phosphor Screens
Use phosphor screens to capture latent images produced by ionizing radiation. Cytiva™ BAS Storage Phosphor Screens emit light upon laser-induced stimulation in proportion to the amount of radioactivity in the sample.
858.00 € - 4735.00 €
Specifications
Quantity 1Each
View More Specs
Products 9
Product Code Brand Product Line Price Quantity & Availability  
Product Code Brand Product Line Price Quantity & Availability  
11360312
View Documents
Cytiva
28-9564-75
BAS-IP MS 2025 E
1496.00 €
Each
In Stock
to see stock.
Add to basket
11350312
View Documents
Cytiva
28-9564-74
BAS-IP MS 2040 E
2384.00 €
Each
In Stock
to see stock.
Add to basket
11370312
View Documents
Cytiva
28-9564-76
BAS-IP MS 3543 E
4735.00 €
Each
In Stock
to see stock.
Add to basket
11310322
View Documents
Cytiva
28-9564-80
BAS-IP SR 0813 N/A N/A N/A
11300322
View Documents
Cytiva
28-9564-79
BAS-IP SR N/A N/A N/A
11380312
View Documents
Cytiva
28-9564-77
SR 2040
3003.00 €
Each
11320322
View Documents
Cytiva
28-9564-81
BAS-IP TR 2040 E
1607.00 €
Each
10232628
View Documents
Cytiva
28-9564-82
TR 20205 E
1066.00 €
Each
11390312
View Documents
Cytiva
28-9564-78
SR 2025
1940.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Requires approximately one-tenth the exposure time of traditional autoradiography film.
  • Ten to one hundred times more sensitive than film, depending on isotope and sample type.
  • Wide linear dynamic range (five orders of magnitude) enables quantitation and visualization of both weak and strong signals in a single exposure.
  • Screens do not require chemicals, darkroom, or other special treatment for use.
  • Each step can be performed at the lab bench under normal lighting conditions and at room temperature.
  • Results are digitized using a storage phosphor imaging system (e.g., Typhoon, Storm, or PhosphorImager) and quantitated using ImageQuant software.
Specifications
Specifications
1Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Cytiva BAS Storage Phosphor Screens