Learn More
MP Biomedicals™ Basal Medium Eagle (Modified), with Hanks' Salts and L-Glutamine, without Sodium Carbonate
Basal Medium Eagle (Modified), with Hanks' Salts and L-Glutamine, without Sodium Carbonate is one of the most commonly used synthetic basal medium for supporting the growth of many different mammalian cells.
Brand:  MP Biomedicals™ 091003122
Product Code. 11440195
  • 45.60 € / 10 L
Explore more special offers
Description
Description
  • Basal Medium Eagle (BME) is one of the most commonly used of all cell culture media.
  • This medium is the result of numerous studies from the late 1950's analyzing the nutrient requirements and other components essential for the growth of cells in culture.
  • Traditionally, BME has been used in studies to measure the growth response of normal (WI-38) and transformed (HeLa) cells in monolayer culture.
  • BME may be used for a broad variety of cell lines when properly supplemented.
  • It has a pH of 6.5 ± 0.5 (w/o sodium bicarb.), 7.3 ± 0.5 (adjusted with sodium bicarb.)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Powder
Bacteria Cell Culture, Mammalian Cell Culture
Suspend 42.5 g of the medium in one liter of distilled or deionized water. Mix well and heat with frequent agitation. Boil for one minute. Dispense and sterilize in an autoclave at 121°C (15 lb. psi) for 15 minutes. Cool to 45 to 50°C and add 5 to 10% sterile defibrinated sheep or rabbit blood if desired.
Polypeptone 10.0, Biotryptase 10.0, Heart Peptone 3.0, Corn Starch 1.0, Sodium Chloride 5.0, Agar 13.5, Final pH 7.3 ±0.2
6.5 to 7.3 ±0.5
10 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.