Learn More
SCHOTT AG Lighting and Imaging™ Base Stand
Base Stand to use with articulated arm (158345) or mounting bracket (120250).
Brand:  SCHOTT AG Lighting and Imaging™ 158340
Product Code. 11368330
  • 91.05 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Includes 3 x M6 thread.
  • Mechanical accessory designed for easy handling.
  • For use with articulating arm (fix flexible light guides or Lightlines) or mounting adapter 120250 for the standalone version with EasyLED Spotlight Plus or KL300LED.
Specifications
Specifications
Base Stand
Articulated Arm (158345) or Mounting Adapter (120250)