missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Universal Valve Lockout Base Clamping Unit

Versatile, rugged, easy to use and lightweight, this Universal Valve Lockout Base Clamping Unit is effective for all types of standard valves. Can also be used to lock out large levers, T-handles and hard-to-secure mechanical devices.

110.00 € - 116.00 €

Specifications

Product Type Valve Lockout Base Clamping Unit
Color Red
Material Reinforced Nylon
Certifications/Compliance OSHA 1910.147
For Use With (Application) Lockout Tagout
View More Specs

Products 2
Product Code Brand HeightMetric Price Quantity & Availability  
Product Code Brand HeightMetric Price Quantity & Availability  
10638245
View Documents
Brady™
050924
110.00 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
10647905
View Documents
Brady™
050899
116.00 €
Each
Estimated Shipment: 09-05-2025
to see stock.
Add to basket
 
Description

Description

  • Base unit only (cable and blocking arm sold separately).
  • Can also be used to lock out large levers, T-handles and hard-to-secure mechanical devices.
  • Open-ended clamp fits over closed rings and wide handles.
  • Made of industrial-grade steel and nylon for extra impact and chemical resistance.
Specifications

Specifications

Valve Lockout Base Clamping Unit
Reinforced Nylon
Lockout Tagout
1 Ea.
Red
OSHA 1910.147
Stainless Steel
1 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Universal Valve Lockout Base Clamping Unit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.