Learn More
BINDER™ Base
Base for use with C 170 and CB 170.
Brand:  BINDER™ 9051-0028
480.19 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 15672206
  • 520.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with CB 170
  • Includes casters
Specifications
Specifications
Base - the base equipped with casters is used for the secure positioning and leveling of a BINDER CO2 incubator
Steel
Casters
BINDER Chambers CB-S 170, CB 170, CBF 170
Stand
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products