missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Basic Agarose Premier

Basic Agarose Premier, Analytical Grade

Brand:  MP Biomedicals™ 11AGAF0500

359.63 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15702442

  • 433.00 € / 500g
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
  • Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9012-36-6
White
Powder
500 g
Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae.
Agarose
Analysis of nucleic acids bygel electrophoresis
Store at Room Temperature (15-30°C).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers