missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Basic Comfort High Lab Chair

Adjustable lab chair with excellent lumbar support for comfortable and healthy seating over a longer period of time.

Brand:  Fisherbrand™ 9651Y-0551-807

206.67 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 17796689

  • 241.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Permanent contact backrest and height-adjustable backrest
  • Large and comfortable seating surface and spacious backrest provide excellent support
  • Reliable body support for laboratory work
  • Easy-care materials, easy to clean
Specifications

Specifications

Chair
620 to 890 mm
GS-mark for tested safety according to DIN 68877
For laboratory work at high laboratory benches
Backrest, seat, gas spring, gas spring cover, foot ring, 5-star foot base, stop&go-castors
120 kg
10 years
Soft stop&go-castors for hard floors
Yes
Yes
High Bench Height
Black
Height adjustable by gas spring
Artificial leather
Polyamide
Gas lift height adjustment
Adjustable from 620 to 890 mm
14 kg
680 mm
Yes
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Basic Comfort High Lab Chair > Type: Chair

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Meets biosafety level S1 and S2, GMP compliant