missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Basic Pole Bag Holder

Basic Pole Bag Holder without wheels, pedal or lid. Bag is very easy to remove and put back. Pole Bag Holder delivered to be assembled.

Brand:  Gilac™ G619071

Product Code. 17948481

  • 162.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pole bag holder without wheels, pedal or lid.
  • Bag very easy to remove and put back.
  • Pole bag holder delivered to be assembled.
Specifications

Specifications

Bag Holder
White
Environment with waste collection needed
Bag Holder
120 L
500 mm
Not Autoclavable
Reusable
Pole bag holder lid + standard 120 L bin bag
HDPE
19.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers