Learn More
Memmert™ Basic Series Water Baths
Corrosion-resistant stainless steel water bath. Memmert™ Basic Series Water Baths feature precise electronics and multiple temperature protection to guarantee the highest level of safety in the laboratory.
695.00 € - 1295.00 €
Specifications
Controller Type Microprocessor PID
Certifications/Compliance CE, VDE, GS
Display Digital LED
For Use With (Application) For research laboratories, food and beverage, temperature control
Material Stainless Steel
View More Specs
Products 6 Description
Description
  • Microprocessor PID temperature controller
  • Digital display
  • Over-temperature protection
  • Integrated digital timer
Specifications
Specifications
Microprocessor PID
Digital LED
Stainless Steel
±0.1°C
50/60 Hz
CE, VDE, GS
For research laboratories, food and beverage, temperature control
95°C
230 V
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Memmert™ Basic Series Water Baths