Learn More
Elma Basket System
Modular basket system for Elmasonic P, Elmasonic Select, EASY
Brand:  Elma 1113387
 View more versions of this product
Product Code. 17808835
  • 159.00 € / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stainless steel with plastic handles
  • Ergonomic handles
  • Autoclavable
  • Bayonet holes in the bottom for mounting holders (e.g. Elma Pin, silicone holder bottom comb)
  • Square openings in the side walls for mounting silicone holders
Specifications
Specifications
Modular Basket System
13.2 lb.
195 x 250 x 90 mm
6.0 kg
Fisherbrand Select. Fisherbrand EASY. Elmasonic EASY. Elmasonic P
Basket System Modular Type 100