Learn More
Julabo™ Bath Adapters
Brand:  Julabo™ 8890009
 View more versions of this product
Product Code. 11954708
  • 186.00 € / Pack of 2
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
CF30, CF31, CF40, CF41 Cryo-compact circulator, F25-HE refrigerated/heating circulator
Adapter
M16 x 1 female to NPT 1/2 in. female