Learn More
Julabo™ Bath Adapters
Brand:  Julabo™ 8890052
 View more versions of this product
Product Code. 13460269
  • 151.00 € / Pack of 2
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Magnum 91, PRESTO™ A30, PRESTO™ A40, PRESTO™ A80, PRESTO™ W40 temperature control/process system
Adapter
M24 x 1/2 in. female to M16 x 1 male