Learn More
Poulten Selfe™ Bath Viscometer Temperature-Controlled
Brand:  Poulten Selfe™ 20405
Product Code. 13161113
  • 6200.00 € / Each
Explore more special offers
Description
Description
4 viscometer positions 25L temperature range 15°C to 60°C 230V 50Hz Poulten Selfe and Lee
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Poulten Selfe™ Bath Viscometer Temperature-Controlled >