missing translation for 'onlineSavingsMsg'
Learn More
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%, Thermo Scientific Chemicals Product Code.: 11344599

Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%, Thermo Scientific Chemicals

Product Code. 11344599
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11344599

Brand: Thermo Scientific Alfa Aesar B23244.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthrolinedisulfonic acid disodium salt hydrate is used as colorimetric reagent for detection of iron. It is widely used for aqueous iron determination without the usage of solvent for extraction. It is used as precursor in the preparation of anionic Europium(III) disulfonated bathophenanthroline tris (dibenzoylmethanate) complex and ruthenium II tris (bathophenanthroline disulfonate). It plays a vital role in the catyltic oxidation of 2-hexanol by making it a water soluble complex with palladium.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52746-49-3
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.48
MDL Number MFCD00149310
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid disodium salt hydrate
PubChem CID 54669730
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 250 mg
Solubility Information Soluble in water.
IUPAC Name disodium 4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzene-1-sulfonate
Formula Weight 536.48 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Bathophenanthrolinedisulfonic acid disodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0440726 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog NumberB23244.06,B23244.MD,B23244.03
Lot Number10237978 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberB23244.06,B23244.MD,B23244.03
Lot Number10243744 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberB23244.06,B23244.MD,B23244.03
Lot Number10242310 Certificate TypeCertificate of Analysis Date22/07/2022 Catalog NumberB23244.06,B23244.MD,B23244.03
Lot Number10240776 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberB23244.06,B23244.MD,B23244.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bathophenanthrolinedisulfonic acid disodium salt hydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.