Learn More
Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90
Safe passive storage of lithium-ion batteries
Brand:  Asecos™ 3725404737264
 View more versions of this product
Product Code. 16389406
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Alert:
For indoor use only
Description
Description
  • Type 90 Safety storage cabinet
  • Tall cabinet with self-closing wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Integrated transport base with removable, optional base cover
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • CE compliant
Specifications
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
615 x 1193 x 2224 mm
Safe passive storage of lithium-ion batteries
IO90.195.120.K1.WDC
4
Powder-coated textured
Sheet Steel
522 x 1050 x 1647 mm
Wing
Bottom collecting sump (sheet steel powder-coated)
2
Grid
Anthracite Gray, Gentian Blue
Storage cabinet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90 > No. of Doors: 2, No. of Shelves: 4