Learn More
TFA Dostmann BatteryCheck Battery Tester
The battery tester BatteryCheck is suitable for the most common battery types (type AAA, AA, C, D), button cells (with 1.2 V / 1.5 V / 3.0 V) and 9 V block batteries. Since it uses the energy of the battery to be tested for the short-term measurement, it does not need its own power supply.
Brand:  TFA Dostmann 98.1126.01
Product Code. 17961662
  • 12.35 € / Each
Explore more special offers
Description
Description
Small, compact and robust and can be used to quick check of batteries and rechargeable batteries: BatteryCheck battery tester. The battery tester is suitable for the most common battery types (type AAA, AA, C, D), button cells (with 1.2 V / 1.5 V / 3.0 V) and 9 V block batteries. Since the BatteryCheck uses the energy of the battery to be tested for the short-term measurement, it does not need its own power supply and it is always ready for use. Perhaps the remaining load is still enough for use in a remote control or a torch.
This way you can use batteries and rechargeable batteries sustainably and help to protect the environment by saving on valuable raw materials. Always dispose of completely discharged batteries and discarded rechargeable batteries responsibly in the collection boxes set up in stores or at an appropriate collection point.
  • Fast and easy check of the power level
  • Suitable for batteries and rechargeable batteries (type AAA, AA, C, D) and button cells of 1.2 V / 1.5 V / 3.0 V and 9 V block batteries
  • Indication of the remaining battery power in volts and charge status with color-coded scale
  • Compact and robust
  • Does not require own power supply
Specifications
Specifications
Battery Tester
Batteries