missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bay 11-7821, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
E2 ubiquitin (Ub) conjugating enzyme inhibitor. Inhibits the conjugation of Ub to a range of E2 enzymes. Inhibits E2 enzyme mediated IκBα phosphorylation and indirectly decreases NF-κb activity. Also reversibly activates MAPK signaling. Induces apoptosis of leukemic T cell and B-cell lymphoma cell lines. Also inhibits activation of NLRP3 inflammasome.
Specifications
Specifications
CAS | 19542-67-7 |
Molecular Formula | C10H9NO2S |
MDL Number | MFCD00712162 |
Quantity | 10 mg |
Synonym | e-3-tosylacrylonitrile, e-3-p-toluenesulfonyl acrylonitrile, unii-4y5g2a4f6o, e-3-4-methylphenyl sulfonylprop-2-enenitrile, 3-p-toluenesulfonyl acrylonitrile, e, 3-4-methylphenylsulfonyl-2-propenenitrile, e-3-4-methylphenylsulfonyl-2-propenenitrile, e 3-4-methylphenyl sulfonyl-2-propenenitrile, 2e-3-4-methylphenyl sulfonyl prop-2-enenitrile, 2-propenenitrile, 3-4-methylphenyl sulfonyl-, 2e |
InChI Key | DOEWDSDBFRHVAP-KRXBUXKQSA-N |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)\C=C\C#N |
IUPAC Name | (2E)-3-(4-methylbenzenesulfonyl)prop-2-enenitrile |
Molecular Weight (g/mol) | 207.25 |
PubChem CID | 5353431 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction