Learn More
Tocris Bioscience™ BAY 876
Potent and selective GLUT1 inhibitor
Brand:  Tocris Bioscience™ 6199/5
Product Code. 18749246
  • 148.00 € / 5mg
Explore more special offers
Description
Description
BAY 876 is a potent and selective GLUT1 inhibitor (IC50 = 2 nM) that displays selectivity for GLUT1 over GLUT2/3/4 (IC50 values are 10.8, 1.67 and 0.29 μM, respectively). BAY 876 induces cell death in hypoxic conditions in vitro and inhibits glucose uptake by Hela-MaTu cells. In ovarian cancer, BAY 876 reduces glycolysis rates and ATP production and inhibits proliferation in vitro and in vivo. This compound is cell permeable and orally bioavailable.
Specifications
Specifications
N4-[1-[(4-Cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-quinolinedicarboxamide
5 mg
98%
1799753-84-6
C24H16F4N6O2