missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Crystal™ ID Systems Accessories

Product Code. p-44609
Click to view available options
Product Type:
Anaerobic Nh inoculum
Calibration Standard
Microbiology Media
Quantity:
10 Pack
Each
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11768912

Brand: BD 245038

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

BBL Crystal MIND Software

  • Interprets the test results and provides specimen identification
  • Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
Codebooks Include Enteric/Nonfermenter, Anaerobe, Neisseria/Haemophilus, Gram-Positive, Rapid Gram-Positive) Supplied in individual hard-copy binder BBL Crystal Panel Viewer
  • For facilitated viewing and recording of chromogenic and fluorogenic reactions
BBL CrystalSpec™ Standards†
  • Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard

†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Anaerobic Nh inoculum
For Use With (Equipment) Crystal™ ID system
Quantity 10 Pack
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.