missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
BD BBL™ Gram Stain Kits
missing translation for 'orderingAttributeHoverText'
Type:
Gram Stain Kit (Stabilized)
Förpackningsstorlek
1kg
Beskrivelse
- Complete stain kits reduce the mess associated with Gram staining procedures
- Each break-resistant solution bottle features a hinged spot cap that makes regulating the flow easy and helps prevent spots and spills Color-coded labels allow quick identification of reagents
- Stabilized Iodine Solution requires no mixing and has a long shelf life
Tekniske data
Tekniske data
| Type | Gram Stain Kit (Stabilized) |
| Quantity | 1 Kit |
| Certifications/Compliance | CE |
| Includes | 250mL gram crystal violet, 250mL iodine, 250mL gram decolorizer and 250mL gram safranin |
| For Use With (Application) | Used to stain microorganisms from cultures or specimens by the differential gram method |
| Stain Type | Gram Stain |
Sikkerhed og håndtering
Produktidentifikator
- Gram Stain Set w/Stabilized Iodine
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H226-Flammable liquid and vapour.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : isopropanol, acetone, ethanol, methanol,isopropanol, ethanol, methanol, C.I. Basic Violet 3, Polyvi
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion