missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ MycoPrep™ Mycobacterial System Digestion/Decontamination Kits

No weighing and no messy mixing

348.00 €

Specifications

Product Type Specimen Collection Kit
Certifications/Compliance CE
For Use With (Application) For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis
Storage Requirements 15°C to 25°C
View More Specs

Products 1
Product Code Brand Volume (Metric) Includes Price Quantity & Availability  
Product Code Brand Volume (Metric) Includes Price Quantity & Availability  
15324564
View Documents
BD
240862
75 mL 10 x 75 mL bottles of NALC-NaOH solution and 5 packets Phosphate Buffer (ph 6.8)
348.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Provide one of the most widely used, CDC-recommended formulations for effective mycobacterial specimen processing
  • Convenient, all-inclusive kits are an alternative to traditional methods of preparing specimen digestion/decontamination reagents
  • Complete with bottles of NALC-NaOH Solution and pre-measured packets of Phosphate Buffer (pH 6.8)
  • Ensure standardization by eliminating the need to weigh, reconstitute or inventory separate ingredients
  • Remain stable for up to 24 hours after activation

Specifications

Specifications

Specimen Collection Kit
CE
For the digestion and decontamination of clinical specimens suspected to contain Mycobacteria, especially Mycobacterium tuberculosis
15°C to 25°C
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 hr.

Product Identifier
  • kit mycoprep

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide

SDS
Documents

Documents

Product Certifications