missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BBP™ Pre-Printed Right-To-Know Chemical Labels with NFPA Diamonds with text ares

Allows printer to produce five-color chemical labels in a single print pass

Brand:  Brady™ 142037

413.34 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 11717174

  • 477.00 € / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Indoor/outdoor-grade vinyl film with permanent adhesive
  • Material withstands exposure to moisture, chemicals and handling
  • Available in multiple sizes and formats
  • Service Temp.: -40° to +82°C (-40° to +180°F)
  • Application Temp.: 18° to 82°C (0° to 180°F)
  • Outdoor Durability: 8 to 10 years

Specifications

Specifications

RoHS 2002/95/EC
Pre-Printed Right-To-Know Chemical Labels for BBP3x Printers
Red, Yellow, Blue, White
Label
NFPA Diamond With Text Area
BBP30, BBP31, BBP33, BBP35, and BBP37 printers
1
170/Pk.
101.6 mm H x 158.8 mm W
-40 to 180°F
0.004 in.
159 x 101 mm (6.25 x 4 in.)
101 mm
101.60 mm
B30-263-595-NFPA
Acrylic, Permanent, Pressure-sensitive
Gloss
Vinyl
For quickly creating NFPA and color bar labels, allows printer to produce 5-color chemical labels in a single print pass, Indoor/outdoor grade material withstands exposure to moisture, chemicals and handling
60 to 50 Gardner Units
8 to 10 Years
4 in. H x 6.25 in. W
Brady Workstation, MarkWare
-40°C to +82°C
0.102 mm
6 in.
4 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.