Learn More
Brady™ BBP12 Label Printer Kit EU with Cutter and Unwinder
The BBP12 Label Printer is a compact, reliable, and affordable entry level thermal transfer benchtop printer ideal for low volume printing on lots of different materials.
Brand:  Brady™ 195966
Product Code. 15717042
  • 102500.00 € / Each
Explore more special offers
Description
Description
The BBP™12 gives you the versatility to print all your basic label types from one machine, saving costs, reducing downtime and simplifying your labeling.
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts for general identification, wire marking, asset tracking and more
  • Specifically designed small-core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Comes with cutter, unwinder, and free Brady™ Workstation Basic Design Software Suite
Specifications
Specifications
Color LCD
8 in.
7.5 in.
10 in.
Not Autoclavable
Monochrome
Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
BBP12
300 dpi
Windows™ 7, Windows™ 8, Windows™ 10
5.1 lb.
Barcode Labeling, Laboratory Labeling, Wire and Cable Labeling
110/240 V
Thermal Transfer
203 mm
191 mm
259 mm
Non-sterile
BBP12 Printer with Pre-installed Cutter, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Brady™ Basic Suite
128 MB (Flash), 64 MB (SDRAM), SD Flash Memory Card Up To 32 GB
101.6 mm/s
Ethernet, Serial, USB
5°C to 40°C
2.3 kg
1 Year
BBP12-EU-U-CUTTER
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products