Learn More
Brady™ BBP12 Label Printer Kit EU with Unwinder, Barcode Reader, and Workstation Scan and Print Suite
This practical kit includes printer, unwinder, barcode reader and Workstation Scan and Print Suite.
Brand:  Brady™ 302670
Product Code. 17135421
  • 1516.62 € / Each
Explore more special offers
Description
Description
The BBP™12 Label Printer packs a lot of punch into one compact tool with its high-speed printing, extended list of labeling options and simple, user-friendly design.
  • Gives versatility to print all basic label types from one machine, saving costs and reducing downtime
  • Over 800 varieties of labels, including external-feed bulk rolls and custom parts for general identification, wire marking, asset tracking and more
  • Specifically designed small core media rolls and ribbons provide fast installation and reduce change-over time
  • Centering ribbon spindle virtually eliminates ribbon wrinkle
  • Compact size won't clutter your workplace or bench
  • Ships with unwinder, barcode reader, and the Brady™ Workstation Scan and Print application for label template design and manually triggered automated label printing
Specifications
Specifications
Color LCD
8 in.
7.5 in.
10 in.
1 in.
Not Autoclavable
Monochrome
Czech, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Kazakh, Lithuanian, Norwegian, Polish, Portuguese, Russian, Slovakian, Spanish, Swedish, Turkish
BBP12
300 dpi
5°C to 40°C
2.3 kg
1 Year
BBP12-SPS-EU
Thermal Transfer
203 mm
191 mm
259 mm
25.4 mm
Non-sterile
Printer, Unwinder, USB Cable, EU Power Cord, Product CD, Quick Start Guide, Xenon 1900HD Barcode Reader, Brady™ Workstation Scan and Print Suite
64 MB
101.6 mm/s
Windows™ 7, Windows™ 8, Windows™ 10
5.1 lb.
Barcode Labeling, Laboratory Labeling, Wire and Cable Labeling
110/240 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products