missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ SER 148 Series Semi-Automatic Solvent Extractor
BIOPHARMA

Operates according to the Randall technique in determining the content of soluble products. Velp Scientifica™ SER 148 Series Semi-Automatic Solvent Extractor can be used to separate a substance from solid and semi-solid samples using immersion, washing and solvent recovery.

Brand:  Velp Scientifica™ F30300240

 View more versions of this product

Product Code. 10787708

  • 8490.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Beats Ordinary Soxhlet Extractor
  • Up to 5 times faster than traditional soxhlet (hot solvent vs. cold solvent)
  • Low solvent consumption (up to 75%)
  • Limited cost per analysis
  • Convenience & Choice
  • Select from 29 on-board empty programs
  • Two models available to match sample load
  • SER 148/3, a 3-position solvent extraction system
  • SER 148/6, a 6-position solvent extraction system
Specifications

Specifications

Solvent Extractor
3 positions, 500 w
1 Ea.
480 mm
30 kg
30 to 100 mL
Stainless Steel
620 mm
480 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.