missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Berkshire™ BCR™ Single-Knit Nylon Full-Finger Cleanroom Glove Liners
Single-knit nylon construction is thin and comfortable, while the full-finger design provides total protection for those with severe glove sensitivities.
Specifications
Construction | Single-Knit |
---|---|
Coating Material | Uncoated |
Color | White |
Disposable | No |
Hand | Ambidextrous |
Description
- Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
- Full-finger design provides total protection for those with severe glove sensitivities
- Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
- Single-knit nylon construction is thin and comfortable
- Ambidextrous design can be used on either hand
- Gloves can be repeatedly laundered for reuse
- Ideal for wet or temperature related processing
- Suitable for use in ISO Class Cleanrooms
Specifications
Single-Knit | |
White | |
Ambidextrous | |
Full | |
Knit |
Uncoated | |
No | |
Nylon | |
Wet/Temperature-Related Processing, Preventing Contact Dermetitis and Perspiration | |
BGL7 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title