missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCX 1470 methanesulfonate, MedChemExpress
MedChemExpress BCX 1470 methanesulfonate inhibits the esterolytic activity of factor D (IC50=96 nM) and C1s (IC50=1.6 nM), 3.4- and 200-fold better, respectively, than that of trypsin.
557.55 € - 876.15 €
Specifications
Chemical Name or Material | BCX 1470 methanesulfonate |
---|---|
CAS | 217099-44-0 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C15H14N2O5S3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16420588
|
Medchem Express
HY-50875 |
5 mg |
557.55 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16408128
|
Medchem Express
HY-50875 |
10 mg |
876.15 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCX 1470 methanesulfonate inhibits the esterolytic activity of factor D (IC50=96 nM) and C1s (IC50=1.6 nM), 3.4- and 200-fold better, respectively, than that of trypsin.Specifications
BCX 1470 methanesulfonate | |
White | |
C15H14N2O5S3 | |
N=C(C1=CC2=CC=C(OC(C3=CC=CS3)=O)C=C2S1)N.O=S(O)(C)=O | |
398.48 | |
Research | |
COVID-19-immunoregulation |
217099-44-0 | |
Research | |
DMSO : ≥ 33.33 mg/mL (83.64 mM) | |
398.48 | |
98.17% | |
Solid |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only