missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Yeast Extract

Gibco™ Bacto™ Dehydrated Culture Media Additive: Yeast Extract is supplied in its dehydrated form for the cultivation of heterotrophic microorganisms.

Brand:  Gibco™ 212750

 View more versions of this product

Product Code. 16279781

  • 150.00 € / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Bacto Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells, derived from primary grown baker's yeast
  • These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
  • Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
  • Bacto Yeast Extract is one of the most complete and versatile fermentation supplements available
  • It is an important ingredient for the microbiological assay of vitamins
  • Yeast extract is also of value in the assay of antibiotics
  • B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
  • Bacto Yeast Extract, Technical, was developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Related yeast extract products include:

  • Gibco Yeast Extract – Developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
  • Difco Yeast Extract UF – Ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g
  • Difco Yeast Extract Low-Dusting (LD) – Designed to limit the amount of dust generated compared to other spray-dried yeast extracts
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Dehydrated
Bottle
Bacto™
Yeast, Animal Origin-Free
Mammalian, Insect, Microbial
Bacterial Media
Formulation of culture media for the cultivation of heterotrophic microorganisms
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3044730 Certificate TypeCertificate of Analysis Date09/01/2025 Catalog Number212750
Lot Number3013745 Certificate TypeCertificate of Analysis Date26/09/2024 Catalog Number212750
Lot Number3013348 Certificate TypeCertificate of Analysis Date26/09/2024 Catalog Number212750
Lot Number3013744 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number212750
Lot Number3013743 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number212750
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Bacto™ Yeast Extract > 500 g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.