missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth
For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium
Brand: BD 249210
This item is not returnable.
View return policy
Description
Specifications
Refrigerate | |
Todd Hewitt Broth | |
Heart, infusion 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L | |
Pail |
Dehydrated | |
Microbiology Media | |
Hemolytic Streptococci growth | |
2 kg |