Learn More
BD Bacto™ Dehydrated Culture Media: Todd Hewitt Broth
For cultivation of group A streptococci, pathogenic microorganisms or as a blood culture medium
Brand:  BD 249220
 View more versions of this product
Product Code. 11798952
  • 2465.00 € / 10kg
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Refrigerate
Todd Hewitt Broth
Heart, infusion 3.1g/L, Neopeptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L
Pail
Dehydrated
Microbiology Media
Hemolytic Streptococci growth
10 kg