Learn More
BD BBL™ Coagulase Plasma, Rabbit, with Citrate
Reconstitutes for use in the Direct Tube Method. BD™ BBL™ Coagulase Plasma, Rabbit, with Citrate is a lyophilized reagent to qualitatively identify and determine the pathogenicity of Staphylococcus aureus.
144.00 € - 429.00 €
Specifications
Species | Rabbit |
---|---|
Color | Light brown |
Content And Storage | 2°C to 8°C |
Form | Lyophilized, Liquid |
Product Type | Coagulase Rabbit Plasma |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11728852
|
BD
240658 |
10 x 3 mL |
144.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11738852
|
BD
240661 |
10 x 15 mL |
429.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Identification of staphylococci is based on microscopic examination, colonial morphology and cultural and biochemical characteristics. Staphylococci associated with acute infection can clot plasma. The most widely used and generally accepted criterion for identification of these pathogenic organisms is based on the presence of the enzyme coagulase.
Staphylococcus aureus produces two types of coagulase, free and bound. Free coagulase is an extracellular enzyme produced when the organism is cultured in broth. Bound coagulase, also known as the clumping factor, remains attached to the cell wall of the organism. In the direct tube test, free coagulase liberated from the cell acts on prothrombin in the coagulase plasma to give a thrombin-like product. This product then acts on fibrinogen to form a fibrin clot.
- Available in 10-tube packs that reconstitute to 3ml or 15ml for use in the Direct Tube Method
- Store reconstituted plasma at 2–8°C for up to 14 day, or aliquot and freeze promptly for up to 30 days
Specifications
Rabbit | |
2°C to 8°C | |
Coagulase Rabbit Plasma | |
Qualitative procedures for the identification of Staphylococcus aureus |
Light brown | |
Lyophilized, Liquid | |
Lyophilized rabbit plasma with 0.85% sodium citrate and 0.85% sodium chloride |
Safety and Handling
missing translation for 'shelfLife' : 1095 Days
Your input is important to us. Please complete this form to provide feedback related to the content on this product.