Learn More
BD BBL™ Crystal™ ID Systems
Miniaturized microbial ID systems use modified conventional, fluorogenic and chromogenic substrates
315.00 € - 394.00 €
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Form | Solid |
Includes | Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations |
Quantity | 20 Pack |
Includes: Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Application) | Price | Quantity & Availability | ||||
11708912
|
BD
245000 |
Enteric/Nonfermenter Kit | Identification of clinically significant aerobic gram negative Enterobacteriaceae isolates and isolated Glucose nonfermenting gram negative Bacilli from Human origin | N/A | |||||
11788922
|
BD
245130 |
Neisseria/Haemophilus ID kit | Identification of clinically significant aerobic and anaerobic gram negative and gram positive bacterial pathogens isolated from Human specimens | N/A | |||||
11718912
|
BD
245010 |
Anaerobe ID Kit | Identification of clinically significant aerobic and anaerobic gram negative and gram positive bacterial pathogens isolated from Human specimens | N/A | |||||
Description
Intended for identification of clinically significant aerobic and anaeorobic gram-negative and gram-positive bacterial pathogens isolated from human specimens. During incubation, the stackable 10-device trays save space and prevent incubator mess. Following incubation, distinct chromogenic and fluorogenic reactions generate a profile number, which is keyed directly into the BBL Crystal MIND Software installed on a laboratory computer.
- Onscreen display identifies the organism, reports on epidemiology, and conveys other pertinent information
- Easy-to-use systems require no pipetting or reagent addition
- Culture colonies are suspended in Crystal Broth; then inoculum suspension is poured directly into the Crystal device
- Wells fill automatically by rocking base
- Lid is snapped onto the device for safety and handling ease
- Enteric/Non-Fermenter (E/NF) Kit performs overnight identification of clinically significant aerobic gram-negative Enterobacteriaceae isolates, as well as frequently isolated glucose nonfermenting gram-negative bacilli from human origin
- Anaerobe ID (ANR) Kit is a four-hour test that identifies clinically significant anaerobes. Separate databases for Schaedler Blood Agar, CDC Anaerobe Blood Agars are included for improved specificity and increased flexibility
- Neisseria/Haemophilus (N/H) ID Kit identifies Neisseria, Haemophilus, Moraxella, Gardnerella vaginalis and other fastidious bacteria in a four-hour test
- Gram-Positive (GP) Kit identifies 121 gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in an overnight test
- Rapid Gram-Positive (RGP) Kit identifies gram-positive bacteria isolated from clinical specimens including both cocci and bacilli in a four-hour test
Specifications
2°C to 8°C | |
Solid | |
Lids, bases, inoculum fluid and incubation trays sufficient for 20 determinations | |
20 Pack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.