Learn More
BD BBL™ Dehydrated Culture Media: Brucella Broth
Dehydrated form for the convenience of the laboratorian and bioprocessing specialist. BD Difco™ BBL™ Dehydrated Culture Media: Brucella Broth is part of a large selection of dehydrated media.
Brand:  BD 211088
Product Code. 11708113
  • 229.00 € / 500g
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dehydrated
  • Culture Media
  • Brucella Broth
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCAM, ISO, USDA
Brucella Broth
Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L
Bottle
Dehydrated
Microbiology Media
Cultivation of Brucella organisms
500 g