missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted)
For antimicrobial susceptibility testing of microorganisms by broth dilution methods. BD Difco™ BBL™ Dehydrated Culture Media: Mueller Hinton II Broth (Cation-Adjusted) is part of a large selection of dehydrated media.
Brand: BD 212322
This item is not returnable.
View return policy
Description
- Dehydrated
- Culture Media
- Mueller Hinton II Broth (Cation-Adjusted)
Specifications
BS12, CLSI, CMPH2, MCM9 | |
Dehydrated | |
Microbiology Media | |
Antimicrobial susceptibility testing of microorganisms by Broth dilution method | |
500 g |
2 to 25°C | |
Mueller Hinton II Broth | |
Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction