Learn More
BD BBL™ MGIT™ Mycobacteria Growth Indicator Tube: Reagents
Offers superior detection time in a user-friendly, fluorescent test format
Brand:  BD 245114
 View more versions of this product
Product Code. 11766924
  • 314.00 € / Pack of 6
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Creates an environment suitable for rapid mycobacterial growth
  • Recommended for testing sputum, stool specimens, body fluids and tissue
  • The tube uses fluorometric technology to aid in the reading of results
  • Under UV light, a positive test emits a vivid orange fluorescent glow at the tube base and at the meniscus; a negative test shows little or no glow
  • The Mycobacteria can be removed from the tube for further testing
  • To use, supplement tube with MGIT OADC Enrichment and MGIT PANTA™ Antibiotic Mixture, inoculate with the prepared specimen and incubate
  • Tubes fit into standard procedures for decontamination and waste disposal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE
2 to 25°C
For the detection and recovery of Mycobacteria, testing Sputum, Stool Specimens, Body Fluids And Tissue
6 Vials