Learn More
BD BBL™ Middlebrook OADC Enrichment
Used as a growth supplement for microbiology research. BD Difco™ BBL™ Middlebrook OADC Enrichment is recommended for isolation and cultivation of mycobacteria and other microbes.
Brand:  BD 211886
 View more versions of this product
Product Code. 11708173
  • 266.00 € / 200mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Dehydrated format
  • Culture media
  • DNR (Latex) free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO/DIS, 11014
Middlebrook OADC Enrichment
Dehydrated culture media
20 mL
2°C to 8°C
Viscous
Tube