Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Gentamicin
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
61.95 €
Specifications
Antimicrobial Agents | Gentamicin |
---|---|
Concentration | 10μg |
Packaging Type | Disc |
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Gentamicin | |
10μg | |
Disc |
Safety and Handling
missing translation for 'shelfLife' : 1460 Days