missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Staphyloslide™ Latex Test Kit

Provides a rapid platform for the identification of Staphylococcal isolates, particularly, Staphylococcus aureus

Brand:  BD 240915

 View more versions of this product

Product Code. 15410937

  • 262.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: BBL Staphyloslide Test Latex; BBL Staphyloslide Control Latex; Test cards, Mixing sticks
This item is not returnable. View return policy

Description

Description

The BD BBL Staphyloslide Latex Test Kit utilizes blue polystyrene latex particles that have been sensitized with fibrinogen and IgG. When Staphylococcal colonies which possess bound coagulase (clumping factor) and/or protein A are mixed with the latex reagent, the latex particles will agglutinate strongly within 20 seconds.

Recommended Storage

Store at 2° to 8°C. Do not freeze.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C Do not freeze
S. aureus
BBL Staphyloslide Test Latex, 2 x 2.5mL; BBL Staphyloslide Control Latex, 2.5mL; Test cards, Mixing sticks
BBL Staphyloslide Latex Test Kit
Staphylococcus aureus Testing
100 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers