missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Taxo™ P Discs (Cartridge)

For presumptive identification of pneumococci, and are intended for use with pure cultures

Brand:  BD 231048

 View more versions of this product

Product Code. 11722924

  • 13.70 € / Pack of 50
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 6mm discs made from high quality absorbent paper
  • Impregnated with approximately 5.0μg of ethylhydrocupreine hydrochloride per disc

Order Info

Cartridges of 50 discs.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ethylhydrocupreine hydrochloride (optochin)
-20°C to +8°C
Cartridge
5μg
Differentiating Pneumococci
50 Discs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For in vitro Diagnostic Use.