Learn More
Bovine serum albumin Stain Buffer, BD
For immunofluorescent staining of single-cell suspensions
Brand: BD Biosciences 554657
242.87 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Alert:
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Description
Useful for dilution and application of fluorescent reagents as well as for suspension, washing, and storage of cells destined for flow cytometric analysis
- Buffered salt solution with neutral pH 7.4
- Supplemented with 0.2% (w/v) bovine serum albumin proteins
- Contains sodium azide which inhibits the potential redistribution of cell surface antigens caused by antibody crosslinking
- Prevents potential loss of fluorescent signal intensities
Flow cytometry, Fluorescence microscopy, Intracellular staining (flow cytometry)

Specifications
Clear | |
Liquid | |
500mL | |
Staining Buffer |
4°C | |
0.2% BSA proteins and 0.09% sodium azide | |
RUO |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research use only