Learn More
Brilliant Stain Buffer, BD
Designed to complement multicolor flow cytometry experiments. Brilliant Stain Buffer is a buffer for the immunofluorescent staining of cells.
Brand:  BD Biosciences 563794
Product Code. 15349374
  • 346.00 € / Each
Edge
Explore more special offers
Description
Description
  • Compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes
  • Research Use Only
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brilliant Stain Buffer, BD > Brilliant Stain buffer; 5mL