Learn More
BD Bromodeoxyuridine (BrdU)
A thymidine analog used in cell proliferation studies
Brand:  BD 550891
Product Code. 10222843
  • 141.00 € / Each
Explore more special offers
Description
Description
BrdU in culture is incorporated into the DNA during DNA synthesis. Cellular incorporation of BrdU can be detected by anti-BrdU specific antibodies following membrane permeabilization through flow cytometry or immunohistochemistry.
  • Free of preservatives; handle under aseptic conditions
  • Provided as five vials, each containing 0.5mL of a 10mg/mL solution diluted in 1X DPBS
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
Bromodeoxyuridine (BrdU)
10 mg/mL
Cellular incorporation of BrdU can be detected by anti-BrdU specific Antibodies following membrane permeabilization through flow cytometry or immunohistochemistry
7.2
-80°C
25 mg