Learn More
BD CBA Human Chemokine Kit
Optimized for analysis of specific chemokines in tissue culture supernatants, EDTA-treated plasma (EDTA-plasma), and serum samples
Brand: BD 552990
1751.59 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes: Reagents for capture and detection of IL-8, RANTES, MIG, MCP-1, and IP-10, wash buffer, assay diluent, cytometer setup reagents
Description
- Uses the sensitivity of amplified fluorescence detection by flow cytometry to measure soluble analytes in a particle-based immunoassay
- Each bead in the kit provides a capture surface for a specific protein and is analogous to an individually coated well in an ELISA plate
- The Capture Bead mixture is in suspension to allow for the detection of multiple analytes in a small volume sample
- The broad dynamic range of fluorescent detection via flow cytometry and the efficient capturing of analytes via suspended particles enable assay to use fewer sample dilutions and to determine the concentration of an unknown analyte in substantially less time (compared to conventional ELISA)
- Can be used to quantitatively measure Interleukin-8 (CXCL8/IL-8), RANTES (CCL5/RANTES), Monokine-induced by Interferon-γ (CXCL9/MIG), Monocyte Chemoattractant Protein-1 (CCL2/MCP-1), and Interferon-γ-induced Protein-10 (CXCL10/IP-10) levels in a single sample
- Provides sufficient reagents for the quantitative analysis of 80 samples
For research use only. Not for use in diagnostic or therapeutic procedures.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Human | |
For analysis of specific chemokines in Tissue Culture supernatants, EDTA-treated Plasma (EDTA-plasma) and Serum samples | |
2°C to 8°C |
IL-8, RANTES, MIG, MCP-1 and IP-10 | |
80 Tests |