Learn More
BD CBA Human Inflammatory Cytokine Kit
Optimized for analysis of specific proteins in tissue culture supernatants, EDTA treated plasma, and serum samples
Brand:  BD 551811
Product Code. 15806708
  • 3970.00 € / 1 set
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Reagents for capture and detection of IL-8, IL-1β, IL-6, IL-10, TNF, and IL-12p70, wash buffer, assay diluent, serum enhancement buffer, cytometer setup reagents
Description
Description
  • Can be used to quantitatively measure interleukin-8 (IL-8), interleukin-1β (IL-1β), interleukin-6 (IL-6), interleukin-10 (IL-10), tumor necrosis factor (TNF), and interleukin-12p70 (IL-12p70) protein levels in a single sample
  • Detection reagent provided in the kit is a mixture of phycoerythrin (PE)–conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
  • Provides sufficient reagents for analysis of 80 samples
For research use only. Not for use in diagnostic or therapeutic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human
For analysis of specific chemokines in Tissue Culture supernatants, EDTA-treated Plasma (EDTA-plasma) and Serum samples
2°C to 8°C
IL-8, IL-1beta, IL-6, IL-10, TNF and IL-12p70
80 Tests