missing translation for 'onlineSavingsMsg'
Learn More

BD Cytofix/Cytoperm Fixation/Permeabilization Solution Kit with GolgiPlug™

For immunofluorescent staining of cytokines

Brand:  BD 555028

Product Code. 15837968

  • 1050.00 € / 1 set
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket

Includes: Fixation/Permeabilization solution (125mL), BD Perm/Wash™ Buffer, 10X concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL), and protein transport inhibitor BD GolgiPlug (containing Brefeldin A)
This item is not returnable. View return policy

Description

Description

  • Offers three kits to simplify fixation and permeabilization of cells
  • For immunofluorescent staining of intracytoplasmic cytokines

All three kits described in this product manual contain Fixation/Permeabilization solution containing 4% paraformaldehyde, and either BD GolgiStop™ protein transport inhibitor containing 99.9% ethanol, or BD GolgiPlug protein transport inhibitor containing 99.9% dimethylsulfoxide and 0.1% brefeldin A. The following warnings and precautions apply to the aforementioned chemicals.
R 36/38: Irritating to eyes and skin
S3: Keep in a cool place
S26: In case of contact with eyes, rinse immediately with water and seek medical advise
S28: After contact with skin, wash immediately with plenty of water
R40: Limited evidence of a carcinogenic effect
S36/37: Wear suitable protective clothing and gloves

Flow Cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

For immunofluorescent staining of cytokines
250 Tests
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only