missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Cytometric Bead Array (CBA) Functional Beads
Color-coded 7.5μm polystyrene beads that can be classified using a flow cytometer
Brand: BD 560039
This item is not returnable.
View return policy
Description
The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.
Specifications
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide | |
Flow Cytometry | |
Functional Bead | |
6000 | |
7.5 μm |
4°C | |
Functional Bead (E7) | |
Polystyrene | |
Flow cytometer | |
0.9 mL |