Learn More
BD Cytometric Bead Array (CBA) Human Angiogenin; Flex Set, Bead C4
For measuring human angiogenin in serum, plasma and cell culture supernatant samples
Brand:  BD 558328
Product Code. 15849308
  • 1425.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Standard, PE Detection Reagent, Capture Bead C4
Description
Description
  • Capture bead and PE detection reagent formulated to 50x concentration
  • Includes lyophilized standard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Angiogenin
1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized)
Serum, Plasma, Cell Culture Supernatant
Flow cytometry, for measuring Human angiogenin
Human
100 Tests
4°C
Human Angiogenin flex set (bead C4)