Learn More
BD Cytometric Bead Array (CBA) Human Eotaxin Flex Set, Bead C7
Sufficient for 100 tests
Brand:  BD 558329
Product Code. 15859308
  • 1030.00 € / Each
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
Includes: Standard, PE Detection Reagent, Capture Bead C7
Description
Description
  • Capture bead and PE detection reagent formulated to 50x concentration
  • Includes lyophilized standard
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Eotaxin
1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized)
Serum, Plasma, Cell Culture Supernatant
Flow Cytometry
Human
100 Tests
4°C
Human eotaxin flex set (bead C7)